Learn More
Abnova™ Human TPRT Partial ORF (NP_055132, 343 a.a. - 415 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_055132 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23590 |
Molecular Weight (g/mol) | 33.77kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16134876
|
Abnova™
H00023590-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16124876
|
Abnova™
H00023590-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is an enzyme that elongates the prenyl side-chain of coenzyme Q, or ubiquinone, one of the key elements in the respiratory chain. The gene product catalyzes the formation of all trans-polyprenyl pyrophosphates from isopentyl diphosphate in the assembly of polyisoprenoid side chains, the first step in coenzyme Q biosynthesis. The protein may be peripherally associated with the inner mitochondrial membrane, though no transit peptide has been definitively identified to date. Defects in this gene are a cause of coenzyme Q10 deficiency. [provided by RefSeq]
Sequence: FPEMNAMIMRRFSLPGDVDRARQYVLQSDGVQQTTYLAQQYCHEAIREISKLRPSPERDALIQLSEIVLTRDKSpecifications
NP_055132 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.77kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COQ1/DPS/MGC70953/RP13-16H11.3/SPS/TPRT/TPT/hDPS1 | |
PDSS1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23590 | |
TPRT (Human) Recombinant Protein (Q01) | |
FPEMNAMIMRRFSLPGDVDRARQYVLQSDGVQQTTYLAQQYCHEAIREISKLRPSPERDALIQLSEIVLTRDK | |
RUO | |
PDSS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |