Learn More
Abnova™ Human TF Partial ORF (AAH59367, 551 a.a. - 650 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH59367 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7018 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16196475
|
Abnova™
H00007018-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16186475
|
Abnova™
H00007018-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a glycoprotein with an approximate molecular weight of 76.5 kDa. It is thought to have been created as a result of an ancient gene duplication event that led to generation of homologous C and N-terminal domains each of which binds one ion of ferric iron. The function of this protein is to transport iron from the intestine, reticuloendothelial system, and liver parenchymal cells to all proliferating cells in the body. This protein may also have a physiologic role as granulocyte/pollen-binding protein (GPBP) involved in the removal of certain organic matter and allergens from serum. [provided by RefSeq]
Sequence: FVKHQTVPQNTGGKNPDPWAKNLNEKDYELLCLDGTRKPVEEYANCHLARAPNHAVVTRKDKEACVHKILRQQQHLFGSNVTDCSGNFCLFRSETKDLLFSpecifications
AAH59367 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781D0156/PRO1557/PRO2086 | |
TF | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
7018 | |
TF (Human) Recombinant Protein (Q01) | |
FVKHQTVPQNTGGKNPDPWAKNLNEKDYELLCLDGTRKPVEEYANCHLARAPNHAVVTRKDKEACVHKILRQQQHLFGSNVTDCSGNFCLFRSETKDLLF | |
RUO | |
TF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |