Learn More
Abnova™ Human SPANXA1 Full-length ORF (AAI48760.1, 1 a.a. - 97 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAI48760.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 30014 |
Molecular Weight (g/mol) | 37.62kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16149312
|
Abnova™
H00030014-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16139312
|
Abnova™
H00030014-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family of cancer/testis-associated genes, which are located in a cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene maps to chromosome X in a head-to-head orientation with SPANX family member A2, which appears to be a duplication of the A1 locus. The protein encoded by this gene targets to the nucleus where it associates with nuclear vacuoles and the redundant nuclear envelope. Based on its association with these poorly characterized regions of the sperm nucleus, this protein provides a biochemical marker to study unique structures in spermatazoa while attempting to further define its role in spermatogenesis. [provided by RefSeq]
Sequence: MDKQSSAGGVKRSVPCDSNEANEMMPETPTGDSDPQPAPKKMKTSESSTILVVRYRRNFKRTSPEELLNDHARENRINPLQMEEEEFMEIMVEIPAKSpecifications
AAI48760.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDKQSSAGGVKRSVPCDSNEANEMMPETPTGDSDPQPAPKKMKTSESSTILVVRYRRNFKRTSPEELLNDHARENRINPLQMEEEEFMEIMVEIPAK | |
RUO | |
SPANXA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
30014 | |
SPANXA1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NAP-X/SPAN-Xa/SPAN-Xb/SPANX/SPANX-A2/SPANX-C/SPANX-D/SPANXA2/SPANXC/SPANXD | |
SPANXA1 | |
Recombinant | |
wheat germ expression system |