missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SNFT Full-length ORF (NP_061134.1, 1 a.a. - 127 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
3655.00 NOK - 5540.00 NOK
Specifications
Accession Number | NP_061134.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55509 |
Molecular Weight (g/mol) | 40.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101513
|
Abnova™
H00055509-P01.25UG |
25 ug |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16191503
|
Abnova™
H00055509-P01.10UG |
10 ug |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This gene encodes a member of the basic leucine zipper protein family. The encoded protein functions as a transcriptional repressor when heterodimerizing with JUN. The protein may play a role in repression of interleukin-2 and matrix metalloproteinase-1 transcription
Sequence: MSQGLPAAGSVLQRSVAAPGNQPQPQPQQQSPEDDDRKVRRREKNRVAAQRSRKKQTQKADKLHEEYESLEQENTMLRREIGKLTEELKHLTEALKEHEKMCPLLLCPMNFVPVPPRPDPVAGCLPRSpecifications
NP_061134.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
JDP1/JUNDM1/SNFT | |
BATF3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55509 | |
SNFT (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
BATF3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |