missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC10A4 Full-length ORF (NP_689892.1, 1 a.a. - 437 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number NP_689892.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 201780
Molecular Weight (g/mol) 72.9kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16189293
View Documents Promotion Details
Abnova™
H00201780-P01.10UG
10 ug
4115.00 NOK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16199293
View Documents Promotion Details
Abnova™
H00201780-P01.25UG
25 ug
6245.00 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence: MDGNDNVTLLFAPLLRDNYTLAPNASSLGPGTDLALAPASSAGPGPGLSLGPGPSFGFSPGPTPTPEPTTSGLAGGAASHGPSPFPRPWAPHALPFWDTPLNHGLNVFVGAALCITMLGLGCTVDVNHFGAHVRRPVGALLAALCQFGLLPLLAFLLALAFKLDEVAAVAVLLCGCCPGGNLSNLMSLLVDGDMNLSIIMTISSTLLALVLMPLCLWIYSWAWINTPIVQLLPLGTVTLTLCSTLIPIGLGVFIRYKYSRVADYIVKVSLWSLLVTLVVLFIMTGTMLGPELLASIPAAVYVIAIFMPLAGYASGYGLATLFHLPPNCKRTVCLETGSQNVQLCTAILKLAFPPQFIGSMYMFPLLYALFQSAEAGIFVLIYKMYGSEMLHKRDPLDEDEDTDISYKKLKEEEMADTSYGTVKAENIIMMETAQTSL
Specifications

Specifications

NP_689892.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
72.9kDa
Glutathione Sepharose 4 Fast Flow
MDGNDNVTLLFAPLLRDNYTLAPNASSLGPGTDLALAPASSAGPGPGLSLGPGPSFGFSPGPTPTPEPTTSGLAGGAASHGPSPFPRPWAPHALPFWDTPLNHGLNVFVGAALCITMLGLGCTVDVNHFGAHVRRPVGALLAALCQFGLLPLLAFLLALAFKLDEVAAVAVLLCGCCPGGNLSNLMSLLVDGDMNLSIIMTISSTLLALVLMPLCLWIYSWAWINTPIVQLLPLGTVTLTLCSTLIPIGLGVFIRYKYSRVADYIVKVSLWSLLVTLVVLFIMTGTMLGPELLASIPAAVYVIAIFMPLAGYASGYGLATLFHLPPNCKRTVCLETGSQNVQLCTAILKLAFPPQFIGSMYMFPLLYALFQSAEAGIFVLIYKMYGSEMLHKRDPLDEDEDTDISYKKLKEEEMADTSYGTVKAENIIMMETAQTSL
RUO
SLC10A4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
201780
SLC10A4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC29802/P4
SLC10A4
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications