Learn More
ACROBiosystems Human Siglec-9 Protein, His Tag
5297.56 NOK - 38899.20 NOK
Specifications
Accession Number | NP_055256.1 |
---|---|
Molecular Weight (g/mol) | 37.9 kDa |
Source | Human Siglec-9, His Tag (SI9-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession # Q9Y336-1). |
Immunogen | Gln 18 - Gly 348 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16458069
|
ACROBiosystems
SI9-H52H4-1MG |
1 mg |
38899.20 NOK
1mg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16448069
|
ACROBiosystems
SI9-H52H4-100UG |
100 μg |
5297.56 NOK
100µg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
siglec-9 (HGMW-approved symbol SIGLEC9) a member of the sialic acid-binding Ig-like lectin (Siglec) family, which belongs to the immunoglobulin superfamily (IgSF). SIGLEC9 shows a high degree of homology to many members of the siglec family, including siglec-7 (80%), siglec-8 (72%), siglec-5 (65%), and CD33 (64%). This high degree of homology is also conserved in the extracellular Ig-like domains. They are characterized by an N-terminal Ig-like V-type domain which mediates sialic acid binding, followed by varying numbers of Ig-like C2-type domains。Siglec-9 with a hydrophobic signal peptide, an N-terminal Ig-likeV-type domain, two Ig-like C2-type domains, a transmembrane region and a cytoplasmic tail.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.9 kDa. The protein migrates as 53-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_055256.1 | |
Human Siglec-9, His Tag (SI9-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession # Q9Y336-1). | |
-20°C | |
Siglec-9 | |
Human | |
25 mM MES, 150 mM NaCl, pH5.5 | |
HEK293 | |
Other Recombinant Proteins |
37.9 kDa | |
Gln 18 - Gly 348 | |
Siglec-9,SIGLEC9,CDw329,CD329 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |