Learn More
Abnova™ Human SGCG Full-length ORF (NP_000222.1, 1 a.a. - 291 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006445-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes gamma-sarcoglycan, one of several sarcolemmal transmembrane glycoproteins that interact with dystrophin. The dystrophin-glycoprotein complex (DGC) spans the sarcolemma and is comprised of dystrophin, syntrophin, alpha- and beta-dystroglycans and sarcoglycans. The DGC provides a structural link between the subsarcolemmal cytoskeleton and the extracellular matrix of muscle cells. Defects in the encoded protein can lead to early onset autosomal recessive muscular dystrophy, in particular limb-girdle muscular dystrophy, type 2C (LGMD2C). [provided by RefSeq]
Sequence: MVREQYTTATEGICIERPENQYVYKIGIYGWRKRCLYLFVLLLLIILVVNLALTIWILKVMWFSPAGMGHLCVTKDGLRLEGESEFLFPLYAKEIHSRVDSSLLLQSTQNVTVNARNSEGEVTGRLKVGPKMVEVQNQQFQINSNDGKPLFTVDEKEVVVGTDKLRVTGPEGALFEHSVETPLVRADPFQDLRLESPTRSLSMDAPRGVHIQAHAGKIEALSQMDILFHSSDGMLVLDAETVCLPKLVQGTWGPSGSSQSLYEICVCPDGKLYLSVAGVSTTCQEHSHICLSpecifications
NP_000222.1 | |
Liquid | |
6445 | |
SGCG (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVREQYTTATEGICIERPENQYVYKIGIYGWRKRCLYLFVLLLLIILVVNLALTIWILKVMWFSPAGMGHLCVTKDGLRLEGESEFLFPLYAKEIHSRVDSSLLLQSTQNVTVNARNSEGEVTGRLKVGPKMVEVQNQQFQINSNDGKPLFTVDEKEVVVGTDKLRVTGPEGALFEHSVETPLVRADPFQDLRLESPTRSLSMDAPRGVHIQAHAGKIEALSQMDILFHSSDGMLVLDAETVCLPKLVQGTWGPSGSSQSLYEICVCPDGKLYLSVAGVSTTCQEHSHICL | |
RUO | |
SGCG | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
A4/DAGA4/DMDA/DMDA1/LGMD2C/MAM/MGC130048/SCARMD2/SCG3/TYPE | |
SGCG | |
Yes | |
wheat germ expression system |