missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SEMA5A Partial ORF (NP_003957, 393 a.a. - 499 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_003957 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9037 |
Molecular Weight (g/mol) | 37.51kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16149785
|
Abnova™
H00009037-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16139785
|
Abnova™
H00009037-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Members of the semaphorin protein family, such as SEMA5A, are involved in axonal guidance during neural development (Adams et al., 1996 [PubMed 8817451]).[supplied by OMIM]
Sequence: PSFMEDNSRFSHVAVDVVQGREALVHIIYLATDYGTIKKVRVPLNQTSSSCLLEEIELFPERRREPIRSLQILHSQSVLFVGLREHVVKIPLKRCQFYRTRSTCIGASpecifications
NP_003957 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.51kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12815/SEMAF/semF | |
SEMA5A | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9037 | |
SEMA5A (Human) Recombinant Protein (Q01) | |
PSFMEDNSRFSHVAVDVVQGREALVHIIYLATDYGTIKKVRVPLNQTSSSCLLEEIELFPERRREPIRSLQILHSQSVLFVGLREHVVKIPLKRCQFYRTRSTCIGA | |
RUO | |
SEMA5A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |