missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human sCD30 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sCD30.

Brand:  Invitrogen™ BMS240TENCE

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15591807

  • 27820.00 NOK / Pack of 10


Estimated Shipment
19-08-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD30
HRP-Conjugate anti-human sCD30 monoclonal antibody
Human sCD30 Standard lyophilized, 200ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

HRP
0.3ng/mL
CD30, sCD30
10 x 96 Tests
Serum
ELISA
25 μL
Lymphoid activation antigen CD30, Ki-1 antigen
Human
Videos
Documents

For Research Use Only