missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RPL3L Full-length ORF (NP_005052.1, 1 a.a. - 407 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_005052.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6123 |
Molecular Weight (g/mol) | 72.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16142348
|
Abnova™
H00006123-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16152348
|
Abnova™
H00006123-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that shares sequence similarity with ribosomal protein L3. The protein belongs to the L3P family of ribosomal proteins. Unlike the ubiquitous expression of ribosomal protein genes, this gene has a tissue-specific pattern of expression, with the highest levels of expression in skeletal muscle and heart. It is not currently known whether the encoded protein is a functional ribosomal protein or whether it has evolved a function that is independent of the ribosome. [provided by RefSeq]
Sequence: MSHRKFSAPRHGHLGFLPHKRSHRHRGKVKTWPRDDPSQPVHLTAFLGYKAGMTHTLREVHRPGLKISKREEVEAVTIVETPPLVVVGVVGYVATPRGLRSFKTIFAEHLSDECRRRFYKDWHKSKKKAFTKACKRWRDTDGKKQLQKDFAAMKKYCKVIRVIVHTQMKLLPFRQKKAHIMEIQLNGGTVAEKVAWAQARLEKQVPVHSVFSQSEVIDVIAVTKGRGVKGVTSRWHTKKLPRKTHKGLRKVACIGAWHPARVGCSIARAGQKGYHHRTELNKKIFRIGRGPHMEDGKLVKNNASTSYDVTAKSITPLGGFPHYGEVNNDFVMLKGCIAGTKKRVITLRKSLLVHHSRQAVENIELKFIDTTSKFGHGRFQTAQEKRAFMGPQKKHLEKETPETSGDLSpecifications
NP_005052.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSHRKFSAPRHGHLGFLPHKRSHRHRGKVKTWPRDDPSQPVHLTAFLGYKAGMTHTLREVHRPGLKISKREEVEAVTIVETPPLVVVGVVGYVATPRGLRSFKTIFAEHLSDECRRRFYKDWHKSKKKAFTKACKRWRDTDGKKQLQKDFAAMKKYCKVIRVIVHTQMKLLPFRQKKAHIMEIQLNGGTVAEKVAWAQARLEKQVPVHSVFSQSEVIDVIAVTKGRGVKGVTSRWHTKKLPRKTHKGLRKVACIGAWHPARVGCSIARAGQKGYHHRTELNKKIFRIGRGPHMEDGKLVKNNASTSYDVTAKSITPLGGFPHYGEVNNDFVMLKGCIAGTKKRVITLRKSLLVHHSRQAVENIELKFIDTTSKFGHGRFQTAQEKRAFMGPQKKHLEKETPETSGDL | |
RUO | |
RPL3L | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6123 | |
RPL3L (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RPL3L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |