Learn More
Abnova™ Human ROBO1 Partial ORF (NP_002932, 491 a.a. - 589 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_002932 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6091 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16144465
|
Abnova™
H00006091-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16134465
|
Abnova™
H00006091-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bilateral symmetric nervous systems have special midline structures that establish a partition between the two mirror image halves. Some axons project toward and across the midline in response to long-range chemoattractants emanating from the midline. The product of this gene is a member of the immunoglobulin gene superfamily and encodes an integral membrane protein that functions in axon guidance and neuronal precursor cell migration. This receptor is activated by SLIT-family proteins, resulting in a repulsive effect on glioma cell guidance in the developing brain. A related gene is located at an adjacent region on chromosome 3. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: DGVLVSTQDSRIKQLENGVLQIRYAKLGDTGRYTCIASTPSGEATWSAYIEVQEFGVPVQPPRPTDPNLIPSAPSKPEVTDVSRNTVTLSWQPNLNSGASpecifications
NP_002932 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DUTT1/FLJ21882/MGC131599/MGC133277/SAX3 | |
ROBO1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6091 | |
ROBO1 (Human) Recombinant Protein (Q01) | |
DGVLVSTQDSRIKQLENGVLQIRYAKLGDTGRYTCIASTPSGEATWSAYIEVQEFGVPVQPPRPTDPNLIPSAPSKPEVTDVSRNTVTLSWQPNLNSGA | |
RUO | |
ROBO1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |