Learn More
Abnova™ Human RMI1 Full-length ORF (AAH64937.1, 1 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00080010-P01.25ug
Additional Details : Weight : 0.00010kg
Description
RMI1 is a component of protein complexes that limit DNA crossover formation via the dissolution of double Holliday junctions (Raynard et al., 2006 [PubMed 16595695]).[supplied by OMIM]
Sequence: MNVTSIALRAETWLLAAWHVKVPPMWLEACINWIQEENNNVNLSQAQMNKQVFEQWLLTDLRDLEHPLLPDGILEIPKGELNGFYALQINSLVDVSQPAHSQIQKLRGKNTTNDLVTAEAQVTPKPWEAKPSRMLMLQLTDGIVQIQGMEYQPIPILHSDLPPGTKILIYGNISFRLGVLLLKPENVKVLGGEVDALLEEYAQEKVLARLIGEPDLVVSVIPNNSNENIPRVTDVLDPALGPSDEELLASLDENDELTANNDTSSERCFTTGSSSNTIPTRQSSFEPEFVISPRPKEEPSNLSIHVMDGELDDFSLEEALLLEETVQKEQMETKELQPLTFNRNADRSIERFSHNPNTTNNFSLTCKNGNNNWSEKNVSEQMTNEDKSFGCPSVRDQNRSIFSVHCNVPLAHDFTNKEKNLETDNKIKQTSSSDSHSLNNKILIERWSTMYRKGIHKFLMKMIVIYRVVLSpecifications
AAH64937.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BLAP75/C9orf76/FLJ12888/RP11-346I8.1 | |
RMI1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
80010 | |
RMI1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNVTSIALRAETWLLAAWHVKVPPMWLEACINWIQEENNNVNLSQAQMNKQVFEQWLLTDLRDLEHPLLPDGILEIPKGELNGFYALQINSLVDVSQPAHSQIQKLRGKNTTNDLVTAEAQVTPKPWEAKPSRMLMLQLTDGIVQIQGMEYQPIPILHSDLPPGTKILIYGNISFRLGVLLLKPENVKVLGGEVDALLEEYAQEKVLARLIGEPDLVVSVIPNNSNENIPRVTDVLDPALGPSDEELLASLDENDELTANNDTSSERCFTTGSSSNTIPTRQSSFEPEFVISPRPKEEPSNLSIHVMDGELDDFSLEEALLLEETVQKEQMETKELQPLTFNRNADRSIERFSHNPNTTNNFSLTCKNGNNNWSEKNVSEQMTNEDKSFGCPSVRDQNRSIFSVHCNVPLAHDFTNKEKNLETDNKIKQTSSSDSHSLNNKILIERWSTMYRKGIHKFLMKMIVIYRVVL | |
RUO | |
RMI1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |