Learn More
Abnova™ Human RAD51L1 Full-length ORF (NP_598193.2, 1 a.a. - 384 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_598193.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5890 |
Molecular Weight (g/mol) | 68.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16142118
|
Abnova™
H00005890-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16152118
|
Abnova™
H00005890-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the RAD51 protein family. RAD51 family members are evolutionarily conserved proteins essential for DNA repair by homologous recombination. This protein has been shown to form a stable heterodimer with the family member RAD51C, which further interacts with the other family members, such as RAD51, XRCC2, and XRCC3. Overexpression of this gene was found to cause cell cycle G1 delay and cell apoptosis, which suggested a role of this protein in sensing DNA damage. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: MGSKKLKRVGLSQELCDRLSRHQILTCQDFLCLSPLELMKVTGLSYRGVHELLCMVSRACAPKMQTAYGIKAQRSADFSPAFLSTTLSALDEALHGGVACGSLTEITGPPGCGKTQFCIMMSILATLPTNMGGLEGAVVYIDTESAFSAERLVEIAESRFPRYFNTEEKLLLTSSKVHLYRELTCDEVLQRIESLEEEIISKGIKLVILDSVASVVRKEFDAQLQGNLKERNKFLAREASSLKYLAEEFSIPVILTNQITTHLSGALASQADLVSPADDLSLSEGTSGSSCVIAALGNTWSHSVNTRLILQYLDSERRQILIAKSPLAPFTSFVYTIKEEGLVLQETTFCSVTQAELNWAPEILPPQPPEQLGLQMCHHTQLIFSpecifications
NP_598193.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGSKKLKRVGLSQELCDRLSRHQILTCQDFLCLSPLELMKVTGLSYRGVHELLCMVSRACAPKMQTAYGIKAQRSADFSPAFLSTTLSALDEALHGGVACGSLTEITGPPGCGKTQFCIMMSILATLPTNMGGLEGAVVYIDTESAFSAERLVEIAESRFPRYFNTEEKLLLTSSKVHLYRELTCDEVLQRIESLEEEIISKGIKLVILDSVASVVRKEFDAQLQGNLKERNKFLAREASSLKYLAEEFSIPVILTNQITTHLSGALASQADLVSPADDLSLSEGTSGSSCVIAALGNTWSHSVNTRLILQYLDSERRQILIAKSPLAPFTSFVYTIKEEGLVLQETTFCSVTQAELNWAPEILPPQPPEQLGLQMCHHTQLIF | |
RUO | |
RAD51L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
5890 | |
RAD51L1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC34245/R51H2/RAD51B/REC2/hREC2 | |
RAD51L1 | |
Recombinant | |
wheat germ expression system |