missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human R-Spondin 1, Recombinant Protein
CHO cells Recombinant Protein
Brand: Invitrogen™ A42586
Additional Details : Weight : 0.02180kg
Description
This gene is a member of the R-spondin family and encodes a secreted activator protein with two cystein-rich, furin-like domains and one thrombospondin type 1 domain. In mice, the protein induces the rapid onset of crypt cell proliferation and increases intestinal epithelial healing, providing a protective effect against chemotherapy-induced adverse effects.Specifications
Q2MKA7 | |
PBS with 0.01% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human R-Spondin 1 | |
CHO cells | |
RUO | |
CRISTIN3; cristin-3; Cysteine-rich and single thrombospondin domain-containing protein 3; FLJ40906; hRspo1; mCristin-3; RGD1565558; Roof plate-specific spondin-1; RP11-566C13.1; RSPO; RSPO1; Rspondin; R-spondin; R-spondin 1; R-spondin homolog; R-spondin homolog (Xenopus laevis); R-spondin-1; thrombospondin type 1 domain containing gene Rspondin | |
Human | |
Recombinant | |
>95% |
Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
284654 | |
50 μg | |
-20°C | |
<1 EU/μg | |
RSPO1 | |
Unconjugated | |
Lyophilized |