missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PURA Partial ORF (NP_005850, 183 a.a. - 292 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number NP_005850
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5813
Molecular Weight (g/mol) 37.84kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16193925
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00005813-Q01.10UG
10 ug
4115.00 NOK
10µg
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16103935
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00005813-Q01.25UG
25 ug
6245.00 NOK
25µg
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia. [provided by RefSeq]

Sequence: TQGQTIALPAQGLIEFRDALAKLIDDYGVEEEPAELPEGTSLTVDNKRFFFDVGSNKYGVFMRVSEVKPTYRNSITVPYKVWAKFGHTFCKYSEETKKIQEKQREKRAAC
Specifications

Specifications

NP_005850
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PUR-ALPHA/PUR1/PURALPHA
PURA
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
5813
PURA (Human) Recombinant Protein (Q01)
TQGQTIALPAQGLIEFRDALAKLIDDYGVEEEPAELPEGTSLTVDNKRFFFDVGSNKYGVFMRVSEVKPTYRNSITVPYKVWAKFGHTFCKYSEETKKIQEKQREKRAAC
RUO
PURA
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates