Learn More
Novus Biologicals™ Human PTGER3 ELISA Kit (Colorimetric)
Human PTGER3 ELISA Kit (Colorimetric) measures PTGER3 in Serum, plasma, cell supernatant, and other biological fluids.
Brand:  Novus Biologicals™ NBP3-00376
Product Code. 18329376
  • 7515.00 NOK / 1 set
Specifications
0.78 to 50 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Human PTGER3 in samples. No significant cross-reactivity or interference between Human PTGER3 and analogues was observed.
ELISA
PTGER3
Cancer, GPCR, Signal Transduction
EP3EP3e, EP3-I, EP3-II, EP3-III, EP3-IV, MGC141828, MGC141829, MGC27302, PGE receptor EP3 subtype, PGE receptor, EP3 subtype, PGE2 receptor EP3 subtype, PGE2-R, prostaglandin E receotor EP3 subtype 3 isoform, prostaglandin E receptor 3 (subtype EP3), prostaglandin E2 receptor EP3 subtype, prostaglandin receptor (PGE-2), Prostanoid EP3 receptor
0.47 ng/mL
Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Human
5733
1 Kit
Storage of components varies. See protocol for specific instructions.
PTGER3