Learn More
Novus Biologicals™ Human PTGER3 ELISA Kit (Colorimetric)
Human PTGER3 ELISA Kit (Colorimetric) measures PTGER3 in Serum, plasma, cell supernatant, and other biological fluids.
Brand: Novus Biologicals™ NBP3-00376
This item is not returnable.
View return policy
Description
Specifications
0.78 to 50 ng/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Human PTGER3 in samples. No significant cross-reactivity or interference between Human PTGER3 and analogues was observed. | |
ELISA | |
PTGER3 | |
Cancer, GPCR, Signal Transduction | |
EP3EP3e, EP3-I, EP3-II, EP3-III, EP3-IV, MGC141828, MGC141829, MGC27302, PGE receptor EP3 subtype, PGE receptor, EP3 subtype, PGE2 receptor EP3 subtype, PGE2-R, prostaglandin E receotor EP3 subtype 3 isoform, prostaglandin E receptor 3 (subtype EP3), prostaglandin E2 receptor EP3 subtype, prostaglandin receptor (PGE-2), Prostanoid EP3 receptor |
0.47 ng/mL | |
Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum | |
Human | |
5733 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
PTGER3 |