missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PSRC1 Full-length ORF (NP_116025.1, 1 a.a. - 333 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_116025.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84722 |
Molecular Weight (g/mol) | 62kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16126053
|
Abnova™
H00084722-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16116053
|
Abnova™
H00084722-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a proline-rich protein. Studies of the related mouse gene suggest that this gene is regulated by p53 and may participate in p53-mediated growth suppression. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]
Sequence: MEDLEEDVRFIVDETLDFGGLSPSDSREEEDITVLVTPEKPLRRGLSHRSDPNAVAPAPQGVRLSLGPLSPEKLEEILDEANRLAAQLEQCALQDRESAGEGLGPRRVKPSPRRETFVLKDSPVRDLLPTVNSLTRSTPSPSSLTPRLRSNDRKGSVRALRATSGKRPSNMKRESPTCNLFPASKSPASSPLTRSTPPVRGRAGPSGRAAASPPTPIRSVLAPQPSTSNSQRLPRPQGAAAKSSSQLPIPSAIPRPASRMPLTSRSVPPGRGALPPDSLSTRKGLPRPSTAGHRVRESGHKVPVSQRLNLPVMGATRSNLQPPRKVAVPGPTRSpecifications
NP_116025.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEDLEEDVRFIVDETLDFGGLSPSDSREEEDITVLVTPEKPLRRGLSHRSDPNAVAPAPQGVRLSLGPLSPEKLEEILDEANRLAAQLEQCALQDRESAGEGLGPRRVKPSPRRETFVLKDSPVRDLLPTVNSLTRSTPSPSSLTPRLRSNDRKGSVRALRATSGKRPSNMKRESPTCNLFPASKSPASSPLTRSTPPVRGRAGPSGRAAASPPTPIRSVLAPQPSTSNSQRLPRPQGAAAKSSSQLPIPSAIPRPASRMPLTSRSVPPGRGALPPDSLSTRKGLPRPSTAGHRVRESGHKVPVSQRLNLPVMGATRSNLQPPRKVAVPGPTR | |
RUO | |
PSRC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84722 | |
PSRC1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DDA3/FP3214/MGC1780/RP11-297O4.2 | |
PSRC1 | |
Recombinant | |
wheat germ expression system |