missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PSG2 Partial ORF (NP_112536, 239 a.a. - 330 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005670-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: PDLPRIHPSYTNYRSGDNLYLSCFANSNPPAQYSWTINGKFQQSGQNLFIPQITTKHSGLYVCSVRNSATGQESSTSLTVKVSASTRIGLLPSpecifications
NP_112536 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.86kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PDLPRIHPSYTNYRSGDNLYLSCFANSNPPAQYSWTINGKFQQSGQNLFIPQITTKHSGLYVCSVRNSATGQESSTSLTVKVSASTRIGLLP | |
RUO | |
PSG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5670 | |
PSG2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CEA/PSBG2/PSG1/PSGGB | |
PSG2 | |
Recombinant | |
wheat germ expression system |