Learn More
Abnova™ Human PPP2R5B Full-length ORF (NP_006235.1, 1 a.a. - 497 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_006235.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5526 |
Molecular Weight (g/mol) | 83.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16141668
|
Abnova™
H00005526-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16131668
|
Abnova™
H00005526-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes a beta isoform of the regulatory subunit B56 subfamily. [provided by RefSeq]
Sequence: METKLPPASTPTSPSSPGLSPVPPPDKVDGFSRRSLRRARPRRSHSSSQFRYQSNQQELTPLPLLKDVPASELHELLSRKLAQCGVMFDFLDCVADLKGKEVKRAALNELVECVGSTRGVLIEPVYPDIIRMISVNIFRTLPPSENPEFDPEEDEPNLEPSWPHLQLVYEFFLRFLESPDFQPSVAKRYVDQKFVLMLLELFDSEDPREREYLKTILHRVYGKFLGLRAYIRKQCNHIFLRFIYEFEHFNGVAELLEILGSIINGFALPLKTEHKQFLVRVLIPLHSVKSLSVFHAQLAYCVVQFLEKDATLTEHVIRGLLKYWPKTCTQKEVMFLGEMEEILDVIEPSQFVKIQEPLFKQVARCVSSPHFQVAERALYFWNNEYILSLIEDNCHTVLPAVFGTLYQVSKEHWNQTIVSLIYNVLKTFMEMNGKLFDELTASYKLEKQQEQQKAQERQELWQGLEELRLRRLQGTQGAKEAPLQRLTPQVAASGGQSSpecifications
NP_006235.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
METKLPPASTPTSPSSPGLSPVPPPDKVDGFSRRSLRRARPRRSHSSSQFRYQSNQQELTPLPLLKDVPASELHELLSRKLAQCGVMFDFLDCVADLKGKEVKRAALNELVECVGSTRGVLIEPVYPDIIRMISVNIFRTLPPSENPEFDPEEDEPNLEPSWPHLQLVYEFFLRFLESPDFQPSVAKRYVDQKFVLMLLELFDSEDPREREYLKTILHRVYGKFLGLRAYIRKQCNHIFLRFIYEFEHFNGVAELLEILGSIINGFALPLKTEHKQFLVRVLIPLHSVKSLSVFHAQLAYCVVQFLEKDATLTEHVIRGLLKYWPKTCTQKEVMFLGEMEEILDVIEPSQFVKIQEPLFKQVARCVSSPHFQVAERALYFWNNEYILSLIEDNCHTVLPAVFGTLYQVSKEHWNQTIVSLIYNVLKTFMEMNGKLFDELTASYKLEKQQEQQKAQERQELWQGLEELRLRRLQGTQGAKEAPLQRLTPQVAASGGQS | |
RUO | |
PPP2R5B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
5526 | |
PPP2R5B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B56B/FLJ35411/PR61B | |
PPP2R5B | |
Recombinant | |
wheat germ expression system |