missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PLEC1 Partial ORF (NP_000436, 4384 a.a. - 4493 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number NP_000436
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5339
Molecular Weight (g/mol) 37.84kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16139181
View Documents Promotion Details
Abnova™
H00005339-Q01.25UG
25 ug
6245.00 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16129181
View Documents Promotion Details
Abnova™
H00005339-Q01.10UG
10 ug
4115.00 NOK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Plectin is a prominent member of plakins or cytolinkers, that are capable of interlinking different elements of the cytoskeleton. Plakins, with their multi-domain structure and enormous size, not only play crucial roles in maintaining cell and tissue integrity and orchestrating dynamic changes in cytoarchitecture and cell shape, but also serve as scaffolding platforms for the assembly, positioning, and regulation of signaling complexes. Plectin is expressed as several protein isoforms in a wide range of cell types and tissues from a single gene located on chromosome 8. The plectin gene locus in mouse on chromosome 15 has been analyzed in detail, revealing a genomic exon-intron organization with well over 40 exons spanning over 62kb and an unusual 5' transcript complexity of plectin isoforms. 11 exons (1-1j) have been identified that alternatively splice directly into a common exon 2 which is the first exon to encode plectin's highly conserved actin binding domain (ABD). Three additional exons (-1, 0a, and 0) splice into an alternative first coding exon (1c), and two additional exons (2a and 3a) are optionally spliced within the exons encoding the acting binding domain (exons 2-8). Analysis of the human locus has identified eight of the eleven alternative 5' exons found in mouse and rat. Furthermore, isoforms lacking the central rod domain encoded by exon 31 have been detected in mouse and rat, and as judged by molecular size, have also been detected in human on the protein level. It has been shown that the short alternative amino-terminal sequences encoded by the different first exons direct the targeting of the various isoforms to distinct subcellular locations. As the expression of specific plectin isoforms was found to be dependent on cell type (tissue) and stage of development it appears that each cell type (tissue) contains a unique set (proportion and composition) of plectin isoforms, as if custom-made for specific requirements of the particular cells. Concordantly, individual isoforms were found to carry out distinct and specific functions.
Sequence: CGFEDPRTKTKMSAAQALKKGWLYYEAGQRFLEVQYLTGGLIEPDTPGRVPLDEALQRGTVDARTAQKLRDVGAYSKYLTCPKTKLKISYKDALDRSMVEEGTGLRLLEA
Specifications

Specifications

NP_000436
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EBS1/EBSO/HD1/PCN/PLEC1b/PLTN
PLEC1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
5339
PLEC1 (Human) Recombinant Protein (Q01)
CGFEDPRTKTKMSAAQALKKGWLYYEAGQRFLEVQYLTGGLIEPDTPGRVPLDEALQRGTVDARTAQKLRDVGAYSKYLTCPKTKLKISYKDALDRSMVEEGTGLRLLEA
RUO
PLEC1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications