missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PIK4CA Partial ORF (NP_002641, 338 a.a. - 447 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_002641 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5297 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16142835
|
Abnova™
H00005297-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16132835
|
Abnova™
H00005297-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a phosphatidylinositol (PI) 4-kinase which catalyzes the first committed step in the biosynthesis of phosphatidylinositol 4,5-bisphosphate. The mammalian PI 4-kinases have been classified into two types, II and III, based on their molecular mass, and modulation by detergent and adenosine. The protein encoded by this gene is a type III enzyme that is not inhibited by adenosine. Two transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Sequence: VPEAIKFLVTWHTIDADAPELSHVLCWAPTDPPTGLSYFSSMYPPHPLTAQYGVKVLRSFPPDAILFYIPQIVQALRYDKMGYVREYILWAASKSQLLAHQFIWNMKTNISpecifications
NP_002641 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ16556/PI4K-ALPHA/PIK4CA/pi4K230 | |
PI4KA | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5297 | |
PIK4CA (Human) Recombinant Protein (Q01) | |
VPEAIKFLVTWHTIDADAPELSHVLCWAPTDPPTGLSYFSSMYPPHPLTAQYGVKVLRSFPPDAILFYIPQIVQALRYDKMGYVREYILWAASKSQLLAHQFIWNMKTNI | |
RUO | |
PI4KA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |