missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PIGB Partial ORF (NP_004846, 80 a.a. - 172 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_004846 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9488 |
Molecular Weight (g/mol) | 35.97kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16130796
|
Abnova™
H00009488-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16140796
|
Abnova™
H00009488-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a transmembrane protein that is located in the endoplasmic reticulum and is involved in GPI-anchor biosynthesis. The glycosylphosphatidylinositol (GPI) anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This gene is thought to encode a member of a family of dolichol-phosphate-mannose (Dol-P-Man) dependent mannosyltransferases. [provided by RefSeq]
Sequence: TSFVPDEYWQSLEVSHHMVFNYGYLTWEWTERLRSYTYPLIFASIYKILHLLGKDSVQLLIWIPRLAQALLSAVADVRLYSLMKQLENQEVARSpecifications
NP_004846 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.97kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC21236 | |
PIGB | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9488 | |
PIGB (Human) Recombinant Protein (Q01) | |
TSFVPDEYWQSLEVSHHMVFNYGYLTWEWTERLRSYTYPLIFASIYKILHLLGKDSVQLLIWIPRLAQALLSAVADVRLYSLMKQLENQEVAR | |
RUO | |
PIGB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |