Learn More
Abnova™ Human PHC2 Full-length ORF (NP_004418.2, 1 a.a. - 323 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_004418.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1912 |
Molecular Weight (g/mol) | 62.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16127297
|
Abnova™
H00001912-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16117297
|
Abnova™
H00001912-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
In Drosophila melanogaster, the 'Polycomb' group (PcG) of genes are part of a cellular memory system that is responsible for the stable inheritance of gene activity. PcG proteins form a large multimeric, chromatin-associated protein complex. The protein encoded by this gene has homology to the Drosophila PcG protein 'polyhomeotic' (Ph) and is known to heterodimerize with EDR1 and colocalize with BMI1 in interphase nuclei of human cells. The specific function in human cells has not yet been determined. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MTSGNGNSASSIAGTAPQNGENKPPQAIVKPQILTHVIEGFVIQEGAEPFPVGRSSLLVGNLKKKYAQGFLPEKLPQQDHTTTTDSEMEEPYLQESKEEGAPLKLKCELCGRVDFAYKFKRSKRFCSMACAKRYNVGCTKRVGLFHSDRSKLQKAGAATHNRRRASKASLPPLTKDTKKQPTGTVPLSVTAALQLTHSQEDSSRCSDNSSYEEPLSPISASSSTSRRRQGQRDLELPDMHMRDLVGMGHHFLPSEPTKWNVEDVYEFIRSLPGCQEIAEEFRAQEIDGQALLLLKEDHLMSAMNIKLGPALKIYARISMLKDSSpecifications
NP_004418.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MTSGNGNSASSIAGTAPQNGENKPPQAIVKPQILTHVIEGFVIQEGAEPFPVGRSSLLVGNLKKKYAQGFLPEKLPQQDHTTTTDSEMEEPYLQESKEEGAPLKLKCELCGRVDFAYKFKRSKRFCSMACAKRYNVGCTKRVGLFHSDRSKLQKAGAATHNRRRASKASLPPLTKDTKKQPTGTVPLSVTAALQLTHSQEDSSRCSDNSSYEEPLSPISASSSTSRRRQGQRDLELPDMHMRDLVGMGHHFLPSEPTKWNVEDVYEFIRSLPGCQEIAEEFRAQEIDGQALLLLKEDHLMSAMNIKLGPALKIYARISMLKDS | |
RUO | |
PHC2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1912 | |
PHC2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EDR2/HPH2/MGC163502/PH2 | |
PHC2 | |
Recombinant | |
wheat germ expression system |