missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PCSK9 (NARC1) His-tag, Recombinant Protein
HEK293 cells Recombinant Protein
Brand: Invitrogen™ A42548
Additional Details : Weight : 0.02180kg
Description
PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.Specifications
Q8NBP7 | |
PBS with 0.01% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human PCSK9 (NARC1) His-tag | |
HEK293 cells | |
RUO | |
AI415265; AI747682; convertase subtilisin; convertase subtilisin/kexin type 9 preproprotein; FH3; HCHOLA3; hypercholesterolemia; LDLCQ1; Narc1; NARC-1; neural apoptosis regulated convertase 1; neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; Pcsk9; proprotein convertase 9; proprotein convertase PC9; proprotein convertase subtilisin/kexin type 9; PSEC0052; subtilisin/kexin-like protease PC9 | |
Human | |
Recombinant | |
>95% |
Control, ELISA, Immunohistochemistry, Western Blot | |
255738 | |
50 μg | |
-20°C | |
<1 EU/μg | |
PCSK9 | |
Unconjugated | |
Lyophilized |