Learn More
Abnova™ Human PCDH11X Partial ORF (NP_061850, 1228 a.a. - 1336 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_061850 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 27328 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16148782
|
Abnova™
H00027328-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16138782
|
Abnova™
H00027328-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its Y homolog, despite divergence leading to coding region changes, is the most closely related cadherin family member. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Transcripts arising from alternative splicing encode isoforms with variable cytoplasmic domains. [provided by RefSeq]
Sequence: AQASALCYSPPLAQAAAISHSSPLPQVIALHRSQAQSSVSLQQGWVQGADGLCSVDQGVQGSATSQFYTMSERLHPSDDSIKVIPLTTFTPRQQARPSRGDSPIMEEHPSpecifications
NP_061850 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PCDH-X/PCDH11/PCDHX/PCDHY | |
PCDH11X | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
27328 | |
PCDH11X (Human) Recombinant Protein (Q01) | |
AQASALCYSPPLAQAAAISHSSPLPQVIALHRSQAQSSVSLQQGWVQGADGLCSVDQGVQGSATSQFYTMSERLHPSDDSIKVIPLTTFTPRQQARPSRGDSPIMEEHP | |
RUO | |
PCDH11X | |
Wheat Germ (in vitro) | |
GST | |
Liquid |