Learn More
Abnova™ Human OSMR Partial ORF (NP_003990.1, 141 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009180-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Oncostatin M is a member of the IL6 family of cytokines. Functional receptors for IL6 family cytokines are multisubunit complexes involving members of the hematopoietin receptor superfamily. Many IL6 cytokines utilize gp130 as a common receptor subunit. OSM binds to the gp130 receptor subunit and, in association with the leukemia inhibitory factor receptor, induces a proliferative response in permissive cells. OSMR is an alternative subunit (for an OSM receptor complex (a heterodimer of gp130 and OSMR) that is activated by OSM but not by LIF [provided by RefSeq]
Sequence: QDSTGQDILFVFPKDKLVEEGTNVTICYVSRNIQNNVSCYLEGKQIHGEQLDPHVTAFNLNSVPFIRNKGTNIYCEASQGNVSEGMKGIVLFVSKVLEEPSpecifications
NP_003990.1 | |
Liquid | |
9180 | |
OSMR (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC150626/MGC150627/MGC75127/OSMRB | |
OSMR | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QDSTGQDILFVFPKDKLVEEGTNVTICYVSRNIQNNVSCYLEGKQIHGEQLDPHVTAFNLNSVPFIRNKGTNIYCEASQGNVSEGMKGIVLFVSKVLEEP | |
RUO | |
OSMR | |
Wheat Germ (in vitro) | |
GST |