missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OSBP Partial ORF (NP_002547.1, 324 a.a. - 407 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_002547.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5007 |
Molecular Weight (g/mol) | 34.98kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16132245
|
Abnova™
H00005007-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16122245
|
Abnova™
H00005007-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Oxysterol binding protein is an intracellular protein that is believed to transport sterols from lysosomes to the nucleus where the sterol down-regulates the genes for the LDL receptor, HMG-CoA reductase, and HMG synthetase [provided by RefSeq]
Sequence: GATVLPANTPGNVGSGKDQCCSGKGDMSDEDDENEFFDAPEIITMPENLGHKRTGSNISGASSDISLDEQYKHQLEETKKEKRTSpecifications
NP_002547.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.98kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
OSBP1 | |
OSBP | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5007 | |
OSBP (Human) Recombinant Protein (Q01) | |
GATVLPANTPGNVGSGKDQCCSGKGDMSDEDDENEFFDAPEIITMPENLGHKRTGSNISGASSDISLDEQYKHQLEETKKEKRT | |
RUO | |
OSBP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |