missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OMD Partial ORF (NP_005005, 274 a.a. - 383 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number NP_005005
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4958
Molecular Weight (g/mol) 37.84kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16132185
View Documents Promotion Details
Abnova™
H00004958-Q01.10UG
10 ug
4115.00 NOK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16142185
View Documents Promotion Details
Abnova™
H00004958-Q01.25UG
25 ug
6245.00 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence: YNIFNLPNIVELSVGHNKLKQAFYIPRNLEHLYLQNNEIEKMNLTVMCPSIDPLHYHHLTYIRVDQNKLKEPISSYIFFCFPHIHTIYYGEQRSTNGQTIQLKTQVFRRF
Specifications

Specifications

NP_005005
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OSAD/SLRR2C
OMD
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
4958
OMD (Human) Recombinant Protein (Q01)
YNIFNLPNIVELSVGHNKLKQAFYIPRNLEHLYLQNNEIEKMNLTVMCPSIDPLHYHHLTYIRVDQNKLKEPISSYIFFCFPHIHTIYYGEQRSTNGQTIQLKTQVFRRF
RUO
OMD
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications