missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NUP54 Full-length ORF (NP_059122.2, 1 a.a. - 507 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_059122.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 53371 |
Molecular Weight (g/mol) | 81.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16150403
|
Abnova™
H00053371-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16140403
|
Abnova™
H00053371-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The nuclear envelope creates distinct nuclear and cytoplasmic compartments in eukaryotic cells. It consists of two concentric membranes perforated by nuclear pores, large protein complexes that form aqueous channels to regulate the flow of macromolecules between the nucleus and the cytoplasm. These complexes are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. This gene encodes a member of the phe-gly (FG) repeat-containing nucleoporin subset. [provided by RefSeq]
Sequence: MAFNFGAPSGTSGTAAATAAPAGGFGGFGTTSTTAGSAFSFSAPTNTGTTGLFGGTQNKGFGFGTGFGTTTGTSTGLGTGLGTGLGFGGFNTQQQQQTTLGGLFSQPTQAPTQSNQLINTASALSAPTLLGDERDAILAKWNQLQAFWGTGKGYFNNNIPPVEFTQENPFCRFKAVGYSCMPSNKDEDGLVVLVFNKKETEIRSQQQQLVESLHKVLGGNQTLTVNVEGTKTLPDDQTEVVIYVVERSPNGTSRRVPATTLYAHFEQANIKTQLQQLGVTLSMTRTELSPAQIKQLLQNPPAGVDPIIWEQAKVDNPDSEKLIPVPMVGFKELLRRLKVQDQMTKQHQTRLDIISEDISELQKNQTTSVAKIAQYKRKLMDLSHRTLQVLIKQEIQRKSGYAIQADEEQLRVQLDTIQGELNAPTQFKGRLNELMSQIRMQNHFGAVRSEERYYIDADLLREIKQHLKQQQEGLSHLISIIKDDLEDIKLVEHGLNETIHIRGGVFSSpecifications
NP_059122.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAFNFGAPSGTSGTAAATAAPAGGFGGFGTTSTTAGSAFSFSAPTNTGTTGLFGGTQNKGFGFGTGFGTTTGTSTGLGTGLGTGLGFGGFNTQQQQQTTLGGLFSQPTQAPTQSNQLINTASALSAPTLLGDERDAILAKWNQLQAFWGTGKGYFNNNIPPVEFTQENPFCRFKAVGYSCMPSNKDEDGLVVLVFNKKETEIRSQQQQLVESLHKVLGGNQTLTVNVEGTKTLPDDQTEVVIYVVERSPNGTSRRVPATTLYAHFEQANIKTQLQQLGVTLSMTRTELSPAQIKQLLQNPPAGVDPIIWEQAKVDNPDSEKLIPVPMVGFKELLRRLKVQDQMTKQHQTRLDIISEDISELQKNQTTSVAKIAQYKRKLMDLSHRTLQVLIKQEIQRKSGYAIQADEEQLRVQLDTIQGELNAPTQFKGRLNELMSQIRMQNHFGAVRSEERYYIDADLLREIKQHLKQQQEGLSHLISIIKDDLEDIKLVEHGLNETIHIRGGVFS | |
RUO | |
NUP54 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
53371 | |
NUP54 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC13407 | |
NUP54 | |
Recombinant | |
wheat germ expression system |