Learn More
Abnova™ Human NOMO2 Partial ORF (NP_775885.1, 1040 a.a. - 1139 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_775885.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 283820 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16152717
|
Abnova™
H00283820-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16142717
|
Abnova™
H00283820-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein originally thought to be related to the collagenase gene family. This gene is one of three highly similar genes in a region of duplication located on the p arm of chromosome 16. These three genes encode closely related proteins that may have the same function. The protein encoded by one of these genes has been identified as part of a protein complex that participates in the Nodal signaling pathway during vertebrate development. Mutations in ABCC6, which is located nearby, rather than mutations in this gene are associated with pseudoxanthoma elasticum (PXE). Two transcripts encoding different isoforms have been described. [provided by RefSeq]
Sequence: NNDIDDVNIIVFRQINQFDLSGNVITSSEYLPTLWVKLYKSENLDNPIQTVSLGQSLFFHFPPLLRDGENYVVLLDSTLPRSQYDYILPQVSFTAVGYHKSpecifications
NP_775885.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Nomo/PM5 | |
NOMO2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
283820 | |
NOMO2 (Human) Recombinant Protein (Q01) | |
NNDIDDVNIIVFRQINQFDLSGNVITSSEYLPTLWVKLYKSENLDNPIQTVSLGQSLFFHFPPLLRDGENYVVLLDSTLPRSQYDYILPQVSFTAVGYHK | |
RUO | |
NOMO2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |