missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NMNAT2 Partial ORF (NP_055854, 208 a.a. - 307 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_055854 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23057 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16136902
|
Abnova™
H00023057-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16126902
|
Abnova™
H00023057-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene product belongs to the nicotinamide mononucleotide adenylyltransferase (NMNAT) enzyme family, members of which catalyze an essential step in NAD (NADP) biosynthetic pathway. Unlike the other human family member, which is localized to the nucleus, and is ubiquitously expressed; this enzyme is cytoplasmic, and is predominantly expressed in the brain. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: CIPGLWNEADMEVIVGDFGIVVVPRDAADTDRIMNHSSILRKYKNNIMVVKDDINHPMSVVSSTKSRLALQHGDGHVVDYLSQPVIDYILKSQLYINASGSpecifications
NP_055854 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C1orf15/KIAA0479/MGC2756/PNAT-2/PNAT2 | |
NMNAT2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23057 | |
NMNAT2 (Human) Recombinant Protein (Q01) | |
CIPGLWNEADMEVIVGDFGIVVVPRDAADTDRIMNHSSILRKYKNNIMVVKDDINHPMSVVSSTKSRLALQHGDGHVVDYLSQPVIDYILKSQLYINASG | |
RUO | |
NMNAT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |