Learn More
Abnova™ Human NFATC3 Full-length ORF (NP_004546.1, 1 a.a. - 1068 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_004546.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4775 |
Molecular Weight (g/mol) | 141.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16110698
|
Abnova™
H00004775-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16120698
|
Abnova™
H00004775-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The product of this gene is a member of the nuclear factors of activated T cells DNA-binding transcription complex. This complex consists of at least two components: a preexisting cytosolic component that translocates to the nucleus upon T cell receptor (TCR) stimulation and an inducible nuclear component. Other members of this family participate to form this complex also. The product of this gene plays a role in the regulation of gene expression in T cells and immature thymocytes. Four transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MTTANCGAHDELDFKLVFGEDGAPAPPPPGSRPADLEPDDCASIYIFNVDPPPSTLTTPLCLPHHGLPSHSSVLSPSFQLQSHKNYEGTCEIPESKYSPLGGPKPFECPSIQITSISPNCHQELDAHEDDLQINDPEREFLERPSRDHLYLPLEPSYRESSLSPSPASSISSRSWFSDASSCESLSHIYDDVDSELNEAAARFTLGSPLTSPGGSPGGCPGEETWHQQYGLGHSLSPRQSPCHSPRSSVTDENWLSPRPASGPSSRPTSPCGKRRHSSAEVCYAGSLSPHHSPVPSPGHSPRGSVTEDTWLNASVHGGSGLGPAVFPFQYCVETDIPLKTRKTSEDQAAILPGKLELCSDDQGSLSPARETSIDDGLGSQYPLKKDSCGDQFLSVPSPFTWSKPKPGHTPIFRTSSLPPLDWPLPAHFGQCELKIEVQPKTHHRAHYETEGSRGAVKASTGGHPVVKLLGYNEKPINLQMFIGTADDRYLRPHAFYQVHRITGKTVATASQEIIIASTKVLEIPLLPENNMSASIDCAGILKLRNSDIELRKGETDIGRKNTRVRLVFRVHIPQPSGKVLSLQIASIPVECSQRSAQELPHIEKYSINSCSVNGGHEMVVTGSNFLPESKIIFLEKGQDGRPQWEVEGKIIREKCQGAHIVLEVPPYHNPAVTAAVQVHFYLCNGKRKKSQSQRFTYTPVLMKQEHREEIDLSSVPSLPVPHPAQTQRPSSDSGCSHDSVLSGQRSLICSIPQTYASMVTSSHLPQLQCRDESVSKEQHMIPSPIVHQPFQVTPTPPVGSSYQPMQTNVVYNGPTCLPINAASSQEFDSVLFQQDATLSGLVNLGCQPLSSIPFHSSNSGSTGHLLAHTPHSVHTLPHLQSMGYHCSNTGQRSLSSPVADQITGQPSSQLQPITYGPSHSGSATTASPAASHPLASSPLSGPPSPQLQPMPYQSPSSGTASSPSPATRMHSGQHSTQAQSTGQGGLSAPSSLICHSLCDPASFPPDGATVSIKPEPEDREPNFATIGLQDITLDDDQFISDLEHQPSGSAEKWPNHSVLSCPAPFWRISpecifications
NP_004546.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
141.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MTTANCGAHDELDFKLVFGEDGAPAPPPPGSRPADLEPDDCASIYIFNVDPPPSTLTTPLCLPHHGLPSHSSVLSPSFQLQSHKNYEGTCEIPESKYSPLGGPKPFECPSIQITSISPNCHQELDAHEDDLQINDPEREFLERPSRDHLYLPLEPSYRESSLSPSPASSISSRSWFSDASSCESLSHIYDDVDSELNEAAARFTLGSPLTSPGGSPGGCPGEETWHQQYGLGHSLSPRQSPCHSPRSSVTDENWLSPRPASGPSSRPTSPCGKRRHSSAEVCYAGSLSPHHSPVPSPGHSPRGSVTEDTWLNASVHGGSGLGPAVFPFQYCVETDIPLKTRKTSEDQAAILPGKLELCSDDQGSLSPARETSIDDGLGSQYPLKKDSCGDQFLSVPSPFTWSKPKPGHTPIFRTSSLPPLDWPLPAHFGQCELKIEVQPKTHHRAHYETEGSRGAVKASTGGHPVVKLLGYNEKPINLQMFIGTADDRYLRPHAFYQVHRITGKTVATASQEIIIASTKVLEIPLLPENNMSASIDCAGILKLRNSDIELRKGETDIGRKNTRVRLVFRVHIPQPSGKVLSLQIASIPVECSQRSAQELPHIEKYSINSCSVNGGHEMVVTGSNFLPESKIIFLEKGQDGRPQWEVEGKIIREKCQGAHIVLEVPPYHNPAVTAAVQVHFYLCNGKRKKSQSQRFTYTPVLMKQEHREEIDLSSVPSLPVPHPAQTQRPSSDSGCSHDSVLSGQRSLICSIPQTYASMVTSSHLPQLQCRDESVSKEQHMIPSPIVHQPFQVTPTPPVGSSYQPMQTNVVYNGPTCLPINAASSQEFDSVLFQQDATLSGLVNLGCQPLSSIPFHSSNSGSTGHLLAHTPHSVHTLPHLQSMGYHCSNTGQRSLSSPVADQITGQPSSQLQPITYGPSHSGSATTASPAASHPLASSPLSGPPSPQLQPMPYQSPSSGTASSPSPATRMHSGQHSTQAQSTGQGGLSAPSSLICHSLCDP | |
RUO | |
NFATC3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
4775 | |
NFATC3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NFAT4/NFATX | |
NFATC3 | |
Recombinant | |
wheat germ expression system |