missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NDRG2 Partial ORF (NP_057334, 1 a.a. - 96 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_057334 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 57447 |
Molecular Weight (g/mol) | 36.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16189096
|
Abnova™
H00057447-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16179096
|
Abnova™
H00057447-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: MAELQEVQITEEKPLLPGQTPEAAKTHSVETPYGSVTFTVYGTPKPKRPAILTYHDVGLNYKSCFQPLFQFEDMQEIIQNFVRVHVDAPGMEEGAPSpecifications
NP_057334 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.3kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781G1938/FLJ25522/KIAA1248/SYLD | |
NDRG2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
57447 | |
NDRG2 (Human) Recombinant Protein (Q01) | |
MAELQEVQITEEKPLLPGQTPEAAKTHSVETPYGSVTFTVYGTPKPKRPAILTYHDVGLNYKSCFQPLFQFEDMQEIIQNFVRVHVDAPGMEEGAP | |
RUO | |
NDRG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |