missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MKKS Full-length ORF (NP_061336.1, 1 a.a. - 570 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008195-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16134678

  • 3602.46 NOK / 10µg

Estimated Shipment: 13-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein with sequence similarity to the chaperonin family. The encoded protein may have a role in protein processing in limb, cardiac and reproductive system development. Mutations in this gene have been observed in patients with Bardet-Biedl syndrome type 6 and McKusick-Kaufman syndrome. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq]

Sequence: MSRLEAKKPSLCKSEPLTTERVRTTLSVLKRIVTSCYGPSGRLKQLHNGFGGYVCTTSQSSALLSHLLVTHPILKILTASIQNHVSSFSDCGLFTAILCCNLIENVQRLGLTPTTVIRLNKHLLSLCISYLKSETCGCRIPVDFSSTQILLCLVRSILTSKPACMLTRKETEHVSALILRAFLLTIPENAEGHIILGKSLIVPLKGQRVIDSTVLPGILIEMSEVQLMRLLPIKKSTALKVALFCTTLSGDTSDTGEGTVVVSYGVSLENAVLDQLLNLGRQLISDHVDLVLCQKVIHPSLKQFLNMHRIIAIDRIGVTLMEPLTKMTGTQPIGSLGSICPNSYGSVKDVCTAKFGSKHFFHLIPNEATICSLLLCNRNDTAWDELKLTCQTALHVLQLTLKEPWALLGGGCTETHLAAYIRHKTHNDPESILKDDECTQTELQLIAEAFCSALESVVGSLEHDGGEILTDMKYGHLWSVQADSPCVANWPDLLSQCGCGLYNSQEELNWSFLRSTRRPFVPQSCLPHEAVGSASNLTLDCLTAKLSGLQVAVETANLILDLSYVIEDKN
Specifications

Specifications

NP_061336.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
88.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BBS6/HMCS/KMS/MKS
MKKS
Yes
wheat germ expression system
Liquid
8195
MKKS (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSRLEAKKPSLCKSEPLTTERVRTTLSVLKRIVTSCYGPSGRLKQLHNGFGGYVCTTSQSSALLSHLLVTHPILKILTASIQNHVSSFSDCGLFTAILCCNLIENVQRLGLTPTTVIRLNKHLLSLCISYLKSETCGCRIPVDFSSTQILLCLVRSILTSKPACMLTRKETEHVSALILRAFLLTIPENAEGHIILGKSLIVPLKGQRVIDSTVLPGILIEMSEVQLMRLLPIKKSTALKVALFCTTLSGDTSDTGEGTVVVSYGVSLENAVLDQLLNLGRQLISDHVDLVLCQKVIHPSLKQFLNMHRIIAIDRIGVTLMEPLTKMTGTQPIGSLGSICPNSYGSVKDVCTAKFGSKHFFHLIPNEATICSLLLCNRNDTAWDELKLTCQTALHVLQLTLKEPWALLGGGCTETHLAAYIRHKTHNDPESILKDDECTQTELQLIAEAFCSALESVVGSLEHDGGEILTDMKYGHLWSVQADSPCVANWPDLLSQCGCGLYNSQEELNWSFLRSTRRPFVPQSCLPHEAVGSASNLTLDCLTAKLSGLQVAVETANLILDLSYVIEDKN
RUO
MKKS
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates