missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MGC2749 Partial ORF (NP_076974, 81 a.a. - 176 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079036-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: FRRIRTLKGKLARQHPEAFSHIPEASFLEEEDEDPIPPSTTTTIATSEQSTGSCDTSPDTVSPSLSPGFEDLSHVQAGSPAINGRSQTDDEEMTGESpecifications
NP_076974 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.3kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FRRIRTLKGKLARQHPEAFSHIPEASFLEEEDEDPIPPSTTTTIATSEQSTGSCDTSPDTVSPSLSPGFEDLSHVQAGSPAINGRSQTDDEEMTGE | |
RUO | |
C19orf50 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
79036 | |
MGC2749 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ25480/MGC2749/MST096/MSTP096 | |
C19orf50 | |
Recombinant | |
wheat germ expression system |