missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MEF2BNB Full-length ORF (AAH04449.1, 1 a.a. - 109 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00729991-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140864

  • 3602.46 NOK / 10µg

Estimated Shipment: 10-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MEEPEMQLKGKKVTDKFTESVYVLANEPSVALYRLQEHVRRSLPELAQHKADMQRWEEQSQGAIYTVEYACSAVKNLVDSSVHFRSVEGLLKQAISIRDHMNASAQGHR
Specifications

Specifications

AAH04449.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MEF2BNB
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
729991
MEF2BNB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MEEPEMQLKGKKVTDKFTESVYVLANEPSVALYRLQEHVRRSLPELAQHKADMQRWEEQSQGAIYTVEYACSAVKNLVDSSVHFRSVEGLLKQAISIRDHMNASAQGHR
RUO
MEF2BNB
Yes
Videos
SDS
Documents
Certificates