missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAN2C1 Partial ORF (NP_006706.2, 258 a.a. - 367 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004123-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140975

  • 6270.00 NOK / 25µg
Estimated Shipment: 29-11-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: HCHIDTAWLWPFKETVRKCARSWVTALQLMERNPEFIFACSQAQQLEWVKSRYPGLYSRIQEFACRGQFVPVGGTWVEMDGNLPSGEAMVRQFLQGQNFFLQEFGKMCSE
Specifications

Specifications

NP_006706.2
Liquid
4123
MAN2C1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686E23167/MAN6A8/MANA/MANA1/MGC87979
MAN2C1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HCHIDTAWLWPFKETVRKCARSWVTALQLMERNPEFIFACSQAQQLEWVKSRYPGLYSRIQEFACRGQFVPVGGTWVEMDGNLPSGEAMVRQFLQGQNFFLQEFGKMCSE
RUO
MAN2C1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers