Learn More
Abnova™ Human MAGEF1 Partial ORF (NP_071432, 198 a.a. - 307 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064110-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This intronless gene encodes a member of the MAGE superfamily. It is ubiquitously expressed in normal tissues and in tumor cells. This gene includes a microsatellite repeat in the coding region. [provided by RefSeq]
Sequence: VQPSKYHFLFGYPKRLIMEDFVQQRYLSYRRVPHTNPPEYEFSWGPRSNLEISKMEVLGFVAKLHKKEPQHWPVQYREALADEADRARAKARAEASMRARASARAGIHLWSpecifications
NP_071432 | |
Liquid | |
64110 | |
MAGEF1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC19617 | |
MAGEF1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VQPSKYHFLFGYPKRLIMEDFVQQRYLSYRRVPHTNPPEYEFSWGPRSNLEISKMEVLGFVAKLHKKEPQHWPVQYREALADEADRARAKARAEASMRARASARAGIHLW | |
RUO | |
MAGEF1 | |
Wheat Germ (in vitro) | |
GST |