missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LGALS4 Partial ORF (NP_006140, 51 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_006140 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3960 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16147381
|
Abnova™
H00003960-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16137381
|
Abnova™
H00003960-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The expression of this gene is restricted to small intestine, colon, and rectum, and it is underexpressed in colorectal cancer. [provided by RefSeq]
Sequence: VVGQDPGSDVAFHFNPRFDGWDKVVFNTLQGGKWGSEERKRSMPFKKGAAFELVFIVLAEHYKVVVNGNPFYEYGHRLPLQMVTHLQVDGDLQLQSINFISpecifications
NP_006140 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GAL4/L36LBP | |
LGALS4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3960 | |
LGALS4 (Human) Recombinant Protein (Q01) | |
VVGQDPGSDVAFHFNPRFDGWDKVVFNTLQGGKWGSEERKRSMPFKKGAAFELVFIVLAEHYKVVVNGNPFYEYGHRLPLQMVTHLQVDGDLQLQSINFI | |
RUO | |
LGALS4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |