missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KLF6 Partial ORF (NP_001291, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_001291 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1316 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16123411
|
Abnova™
H00001316-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16113411
|
Abnova™
H00001316-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the Kruppel-like family of transcription factors. The zinc finger protein is a transcriptional activator, and functions as a tumor suppressor. Multiple transcript variants encoding different isoforms have been found for this gene, some of which are implicated in carcinogenesis. [provided by RefSeq]
Sequence: MDVLPMCSIFQELQIVHETGYFSALPSLEEYWQQTCLELERYLQSEPCYVSASEIKFDSQEDLWTKIILAREKKEESELKISSSPPEDTLISPSFCYNLESpecifications
NP_001291 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCD1/COPEB/CPBP/DKFZp686N0199/GBF/PAC1/ST12/ZF9 | |
KLF6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1316 | |
KLF6 (Human) Recombinant Protein (Q01) | |
MDVLPMCSIFQELQIVHETGYFSALPSLEEYWQQTCLELERYLQSEPCYVSASEIKFDSQEDLWTKIILAREKKEESELKISSSPPEDTLISPSFCYNLE | |
RUO | |
KLF6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |