Learn More
Abnova™ Human KLC3 Full-length ORF (AAH73841.1, 1 a.a. - 503 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00147700-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the kinesin light chain gene family. Kinesins are molecular motors involved in the transport of cargo along microtubules, and are composed of two kinesin heavy chain (KHC) and two kinesin light chain (KLC) molecules. KLCs are thought to typically be involved in binding cargo and regulating kinesin activity. In the rat, a protein similar to this gene product is expressed in post-meiotic spermatids, where it associates with structural components of sperm tails and mitochondria. [provided by RefSeq]
Sequence: MSVQVAAPGSAGLGPERLSPEELVRQTRQVVQGLEALRAEHHGLAGHLAEALAGQGPAAGLEMLEEKQQVVSHSLEAIELGLGEAQVLLALSAHVGALEAEKQRLRSQARRLAQENVWLREELEETQRRLRASEESVAQLEEEKRHLEFLGQLRQYDPPAESQSESPPRRDSLASLFPSEEEERKGPEAAGAAAAQQGGYEIPARLRTLHNLVIQYAGQGRYEVAVPLCRQALEDLERSSGHCHPDVATMLNILALVYRDQNKYKEATDLLHDALQIREQTLGPEHPAVAATLNNLAVLYGKRGRYREAEPLCQRALEIREKVLGADHPDVAKQLNNLALLCQNQGKFEDVERHYARALSIYEALGGPHDPNVAKTKNNLASAYLKQNKYQQAEELYKEILHKEDLPAPLGAPNTGTAGDAEQALRRSSSLSKIRESIRRGSEKLVSRLRGEAAAGAAGMKRAMSLNTLNVDAPRAPGTQFPSWHLDKAPRTLSASTQDLSPHSpecifications
AAH73841.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KLC2/KLC2L/KLCt/KNS2B | |
KLC3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
147700 | |
KLC3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSVQVAAPGSAGLGPERLSPEELVRQTRQVVQGLEALRAEHHGLAGHLAEALAGQGPAAGLEMLEEKQQVVSHSLEAIELGLGEAQVLLALSAHVGALEAEKQRLRSQARRLAQENVWLREELEETQRRLRASEESVAQLEEEKRHLEFLGQLRQYDPPAESQSESPPRRDSLASLFPSEEEERKGPEAAGAAAAQQGGYEIPARLRTLHNLVIQYAGQGRYEVAVPLCRQALEDLERSSGHCHPDVATMLNILALVYRDQNKYKEATDLLHDALQIREQTLGPEHPAVAATLNNLAVLYGKRGRYREAEPLCQRALEIREKVLGADHPDVAKQLNNLALLCQNQGKFEDVERHYARALSIYEALGGPHDPNVAKTKNNLASAYLKQNKYQQAEELYKEILHKEDLPAPLGAPNTGTAGDAEQALRRSSSLSKIRESIRRGSEKLVSRLRGEAAAGAAGMKRAMSLNTLNVDAPRAPGTQFPSWHLDKAPRTLSASTQDLSPH | |
RUO | |
KLC3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |