missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIAA0527 Partial ORF (XP_171054, 284 a.a. - 383 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026032-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135276

  • 6270.00 NOK / 25µg
Estimated Shipment: 06-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: TTVCSKGSGEQQIMRAVDVRIESNPVPGGTYSALCIKDEEKPCGDPPSFPHTILQGRTGLEMGDELLYVCAPGHIMGHRETAFTLLCNSCGEWYGLVQAC
Specifications

Specifications

XP_171054
Liquid
26032
KIAA0527 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0527
SUSD5
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TTVCSKGSGEQQIMRAVDVRIESNPVPGGTYSALCIKDEEKPCGDPPSFPHTILQGRTGLEMGDELLYVCAPGHIMGHRETAFTLLCNSCGEWYGLVQAC
RUO
SUSD5
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers