Learn More
Abnova™ Human ITGAE Partial ORF (NP_002199, 57 a.a. - 171 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_002199 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3682 |
Molecular Weight (g/mol) | 38.39kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16150285
|
Abnova™
H00003682-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16140285
|
Abnova™
H00003682-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes an I-domain-containing alpha integrin that undergoes post-translational cleavage in the extracellular domain, yielding disulfide-linked heavy and light chains. In combination with the beta 7 integrin, this protein forms the E-cadherin binding integrin known as the human mucosal lymphocyte-1 antigen. This protein is preferentially expressed in human intestinal intraepithelial lymphocytes (IEL), and in addition to a role in adhesion, it may serve as an accessory molecule for IEL activation. [provided by RefSeq]
Sequence: SPRTKRTPGPLHRCSLVQDEILCHPVEHVPIPKGRHRGVTVVRSHHGVLICIQVLVRRPHSLSSELTGTCSLLGPDLRPQAQANFFDLENLLDPDARVDTGDCYSNKEGGGEDDVSpecifications
NP_002199 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.39kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD103/HUMINAE/MGC141996 | |
ITGAE | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3682 | |
ITGAE (Human) Recombinant Protein (Q01) | |
SPRTKRTPGPLHRCSLVQDEILCHPVEHVPIPKGRHRGVTVVRSHHGVLICIQVLVRRPHSLSSELTGTCSLLGPDLRPQAQANFFDLENLLDPDARVDTGDCYSNKEGGGEDDV | |
RUO | |
ITGAE | |
Wheat Germ (in vitro) | |
GST | |
Liquid |