Learn More
Abnova™ Human IL1RL2 Partial ORF (NP_003845, 20 a.a. - 118 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_003845 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8808 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16199315
|
Abnova™
H00008808-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16189315
|
Abnova™
H00008808-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the interleukin 1 receptor family. An experiment with transient gene expression demonstrated that this receptor was incapable of binding to interleukin 1 alpha and interleukin 1 beta with high affinity. This gene and four other interleukin 1 receptor family genes, including interleukin 1 receptor, type I (IL1R1), interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 1 (IL1RL1), and interleukin 18 receptor 1 (IL18R1), form a cytokine receptor gene cluster in a region mapped to chromosome 2q12. [provided by RefSeq]
Sequence: DGCKDIFMKNEILSASQPFAFNCTFPPITSGEVSVTWYKNSSKIPVSKIIQSRIHQDETWILFLPMEWGDSGVYQCVIKGRDSCHRIHVNLTVFEKHWCSpecifications
NP_003845 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL1R-rp2/IL1RRP2 | |
IL1RL2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8808 | |
IL1RL2 (Human) Recombinant Protein (Q01) | |
DGCKDIFMKNEILSASQPFAFNCTFPPITSGEVSVTWYKNSSKIPVSKIIQSRIHQDETWILFLPMEWGDSGVYQCVIKGRDSCHRIHVNLTVFEKHWC | |
RUO | |
IL1RL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |