Learn More
R&D Systems™ Human IL-1 beta/IL-1F2 High Sensitivity Magnetic Luminex™ Performance Assay
Product Code. 18462260
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 18462260
Brand: R&D Systems™ LHSCM201
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
  • Magnetic format allows for easier wash steps
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
  • Requires a small sample volume (<50 μL)
  • Run your assay in just 3-5 hours
  • Mass-calibrated standards for consistent results with every new lot of material
  • Enough reagents for one 96-well plate
TRUSTED_SUSTAINABILITY
Specifications
Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target IL-1 beta/IL-1F2
Bead Type Magnetic
Assay Sensitivity 0.18pg/mL
Assay Range 0.29 - 1200pg/mL
Gene ID (Entrez) 3553
Gene Symbol IL1B
Sample Type Serum, Plasma (EDTA, Heparin)
Sample Volume Serum (50μL),EDTA Plasma (50μL),Heparin Plasma (50μL)
Synonym catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Kit Contents
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Content And Storage Store the unopened product at 2° to 8°C. Do not use past expiration date.