missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IGSF1 Partial ORF (NP_001546, 220 a.a. - 310 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_001546 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3547 |
Molecular Weight (g/mol) | 35.75kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16126721
|
Abnova™
H00003547-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16116721
|
Abnova™
H00003547-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Members of the immunoglobulin (Ig) superfamily (see MIM 147100), which includes IGSF1, have a variety of functions, but all appear to play a role in cell recognition and the regulation of cell behavior.[supplied by OMIM]
Sequence: VVAGLYPKPTLTAHPGPIMAPGESLNLRCQGPIYGMTFALMRVEDLEKSFYHKKTIKNEANFFFQSLKIQDTGHYLCFYYDASYRGSLLSDSpecifications
NP_001546 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IGCD1/IGDC1/INHBP/KIAA0364/MGC75490/PGSF2 | |
IGSF1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3547 | |
IGSF1 (Human) Recombinant Protein (Q01) | |
VVAGLYPKPTLTAHPGPIMAPGESLNLRCQGPIYGMTFALMRVEDLEKSFYHKKTIKNEANFFFQSLKIQDTGHYLCFYYDASYRGSLLSD | |
RUO | |
IGSF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |