missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IgM Recombinant Isotype Control (SAA0185)

Product Code. 30132211
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132211 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30132211 Supplier Invitrogen™ Supplier No. MA555117

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Recombinant Monoclonal Isotype Control

Human IgM Isotype Control for Flow

IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgM
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone SAA0185
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH
Host Species Human
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Target Species Not Applicable
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Isotype Control
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.