missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IgA Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IgA.

Brand:  Invitrogen™ BMS2096TENCE

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15591637

  • 27830.00 NOK / Pack of 10


Estimated Shipment
19-08-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgA

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
0.03ng/mL
2°C to 8°C
Immunoglobulin A
Human
ELISA
20 μL (1:10,000 prediluted)
IgA
10 x 96 Tests
Serum
Videos
Documents

For Research Use Only