missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HSH2D Partial ORF (NP_116244.1, 251 a.a. - 352 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_116244.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84941 |
Molecular Weight (g/mol) | 36.96kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16150967
|
Abnova™
H00084941-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16140967
|
Abnova™
H00084941-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
T-cell activation requires 2 signals: recognition of antigen by the T-cell receptor (see TCR; MIM 186880) and a costimulatory signal provided primarily by CD28 (MIM 186760) in naive T cells. HSH2 is a target of both of these signaling pathways (Greene et al., 2003 [PubMed 12960172]).[supplied by OMIM]
Sequence: SQDHSGDPTSGDRGYTDPCVATSLKSPSQPQAPKDRKVPTRKAERSVSCIEVTPGDRSWHQMVVRALSSQESKPEHQGLAEPENDQLPEEYQQPPPFAPGYCSpecifications
NP_116244.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALX/FLJ14886/HSH2 | |
HSH2D | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
84941 | |
HSH2D (Human) Recombinant Protein (Q01) | |
SQDHSGDPTSGDRGYTDPCVATSLKSPSQPQAPKDRKVPTRKAERSVSCIEVTPGDRSWHQMVVRALSSQESKPEHQGLAEPENDQLPEEYQQPPPFAPGYC | |
RUO | |
HSH2D | |
Wheat Germ (in vitro) | |
GST | |
Liquid |