missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HSDL2 Full-length ORF (NP_115679.2, 1 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_115679.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84263 |
Molecular Weight (g/mol) | 71.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16125753
|
Abnova™
H00084263-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16115753
|
Abnova™
H00084263-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: MLPNTGRLAGCTVFITGASRGIGKAIALKAAKDGANIVIAAKTAQPHPKLLGTIYTAAEEIEAVGGKALPCIVDVRDEQQISAAVEKAIKKFGGIDILVNNASAISLTNTLDTPTKRLDLMMNVNTRGTYLASKACIPYLKKSKVAHILNISPPLNLNPVWFKQHCAYTIAKYGMSMYVLGMAEEFKGEIAVNALWPKTAIHTAAMDMLGGPGIESQCRKVDIIADAAYSIFQKPKSFTGNFVIDENILKEEGIENFDVYAIKPGHPLQPDFFLDEYPEAVSKKVESTGAVPEFKEEKLQLQPKPRSGAVEETFRIVKDSLSDDVVKATQAIYLFELSGEDGGTWFLDLKSKGGNVGYGEPSDQADVVMSMTTDDFVKMFSGKLKPTMAFMSGKLKIKGNMALAIKLEKLMNQMNARLSpecifications
NP_115679.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLPNTGRLAGCTVFITGASRGIGKAIALKAAKDGANIVIAAKTAQPHPKLLGTIYTAAEEIEAVGGKALPCIVDVRDEQQISAAVEKAIKKFGGIDILVNNASAISLTNTLDTPTKRLDLMMNVNTRGTYLASKACIPYLKKSKVAHILNISPPLNLNPVWFKQHCAYTIAKYGMSMYVLGMAEEFKGEIAVNALWPKTAIHTAAMDMLGGPGIESQCRKVDIIADAAYSIFQKPKSFTGNFVIDENILKEEGIENFDVYAIKPGHPLQPDFFLDEYPEAVSKKVESTGAVPEFKEEKLQLQPKPRSGAVEETFRIVKDSLSDDVVKATQAIYLFELSGEDGGTWFLDLKSKGGNVGYGEPSDQADVVMSMTTDDFVKMFSGKLKPTMAFMSGKLKIKGNMALAIKLEKLMNQMNARL | |
RUO | |
HSDL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84263 | |
HSDL2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C9orf99/FLJ25855/MGC10940/SDR13C1 | |
HSDL2 | |
Recombinant | |
wheat germ expression system |